missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Heavy-duty Shoe Covers
These are lint free PE-PP non-skid shoe covers that protect against dirt and grime in heavy-duty conditions.
$79.50 - $79.50
Specifications
| Material | Laminated Polypropylene, Non-woven Spunbond Polypropylene |
|---|---|
| Latex Free | No |
| Coating | Polypropylene |
| Color | Blue |
| Disposable | Single-use |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
12-896-0024XL
|
Fisherbrand™
128960024XL |
X-Large |
Case of 300 Each for $79.50
|
|
|||||
Description
- Heavy-duty PE
- Disposable
- Non-Skid
- Protect against dirt and grime in heavy-duty conditions.
Specifications
| Laminated Polypropylene, Non-woven Spunbond Polypropylene | |
| Polypropylene | |
| Single-use | |
| Elastic Ankle | |
| Fluid |
| No | |
| Blue | |
| Non-sterile | |
| Anti-skid | |
| Shoe Covers |