missing translation for 'onlineSavingsMsg'
Learn More

Keystone Safety Heavy-Duty Polypropylene Coveralls, CVL-NW-HD-E

Catalog No. 19055278
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6X-Large
Color:
Blue
White
Catalog No. Size Color
19055278 Medium White
19055279 Large White
19055280 X-Large White
19055271 X-Large Blue
19055281 2X-Large White
19055282 3X-Large White
19055276 4X-Large White
19055274 4X-Large Blue
19070136 6X-Large White
9 options

Catalog No. 19055278

Supplier: Keystone Safety CVLNWHDEMEDWHITE

Only null left

Heavyweight polypropylene coveralls are made of breathable fabric that provides economical protection against dry particulates, dirt, and grime. Available in blue or white.

  • Heavyweight polypropylene coveralls
  • Breathable fabric provides economical protection
  • Elastic wrists and ankles
  • Low cost solution to protect against dry particulates, dirt, and grime
  • 100% latex free

Specifications

Product Type General Purpose Coveralls
Size Medium
Gender Unisex
Material Polypropylene (Heavy-duty)
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Particle
Certifications/Compliance Cleanroom Class 7
Latex Free Yes
Model CVL-NW-HD-E-LRG
No. of Pockets None
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Quantity 25/Cs.
Show More Show Less