missing translation for 'onlineSavingsMsg'
Learn More
Learn More
International Enviroguard™ Heavy Duty 3407 CPE Shoe Covers
Description
Disposable shoe covers are built to last even in the most rugged environments. Heavy-duty CPE shoe covers are made of coated polyethylene to help protect against non-hazardous particles, liquid splash, and shallow immersion. Diamond pattern bottom offers skid resistance, enhanced traction, and durability.
- Water resistant and come with a strong elastic closure
- One size fits most; Fits up to a men's size 12 1/2 shoe
- Elastic cuffs for a secure fit around the ankles
- Can be worn until damaged or contaminated
- Available in both White and Blue
- Possible applications include construction, healthcare, cleanrooms, food processing, contract work, pharmaceuticals, moving services, and more
Specifications
Specifications
| Size | Universal |
| Material | CPE |
| Latex Free | Yes |
| Color | White |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Closure Type | Elastic Ankle |
| Sole Type | Skid-resistant |
| Seam Type | Molded |
| Packaging Quantity | 300/Cs. |
| Show More |