missing translation for 'onlineSavingsMsg'
Learn More

Heavy-Duty Batteries

Catalog No. p-2365833
Change view
Click to view available options
Battery Type:
AA
D
12 V
Quantity:
1 Ea.
6/Pk. (Shrink-wrapped packs)
8/Pk. (Shrink-wrapped packs)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Battery Type Quantity
S43409 12 V 1 Ea.
S43923H D 6/Pk. (Shrink-wrapped packs)
S43921H AA 8/Pk. (Shrink-wrapped packs)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S43409 Supplier Customer Channels Group (Ccg) Supplier No. 926
Only null left

For low-drain devices or devices that are used infrequently.

Use heavy-duty batteries for long-term applications like:

  • Remote controls
  • Clocks
  • Smoke alarms
  • Radios
  • Flashlights

Specifications

Product Type Heavy-duty Battery
Battery Type 12 V
Cell Type Screw Terminal
Certifications/Compliance EPA
For Use With (Application) For low drain devices or devices that are used infrequently
For Use With (Equipment) For remote controls, clocks, smoke alarms, radios, flashlights
Mounting Type Screw
Quantity 1 Ea.
Voltage 12 V