Learn More
Research Products Heating Block Module for 1.5ml Microtubes, 24 Tube Capacity
Supplier: Research Products Corporation RPI110040
These precision machined solid aluminum blocks are meant to accommodate micro-centrifuge tubes, microplates and test tubes. Anodized blocks resist corrosion and feature alpha/numerical labeled rows and columns for easy vessel identification. Each block has a well for a thermometer (sold separately) and a block extractor hole.
Block Modules can be used in either an analog or digital dry bath incubator.