missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synthware™ Heating Transfer Block

Specifications
Specifications
| Color | Silver |
| Capacity (Metric) | 3, 5 mL |
| Material | Aluminum |
| For Use With (Application) | Organic Synthesis |
| For Use With (Equipment) | Holds 3 and 5 mL heavywall reaction vials, 4 Craig crystallization tubes, 1 standard thermometer |