Learn More
Chemglass Inc 500mL Glas-Col Heating Mantle, Series O, Cylindrical, 4 3/16in ID x 4in Depth, 250 Watts
Catalog No. CG1000710
Encompass
500mL Glas-Col Heating Mantle, Series O, Cylindrical, 4 3/16in ID x 4in Depth, 250 Watts. This Glas-Col Series O mantle is used with a round bottom cylindrical reaction vessel, CG-1920 and CG-1922, featuring a Duran flange. The mantle covers the bottom and sides of the vessel leaving enough room under the flange to safely remove the reaction vessel clamp while the mantle is in place. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector. The 10,000mL size has two circuits at the listed wattage.
Catalog No. CG1000710 Supplier Chemglass Inc Supplier No. CG1000710
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
500mL Glas-Col Heating Mantle, Series O, Cylindrical, 4 3/16in ID x 4in Depth, 250 Watts. This Glas-Col Series O mantle is used with a round bottom cylindrical reaction vessel, CG-1920 and CG-1922, featuring a Duran flange. The mantle covers the bottom and sides of the vessel leaving enough room under the flange to safely remove the reaction vessel clamp while the mantle is in place. Mantle is glass fabric, 115 volts, and is supplied with a separable 4 foot, 2 wire extension cord with locking connector. The 10,000mL size has two circuits at the listed wattage.