missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Heating Blocks for Microtube Shaker

Catalog No. 14223094
Change view
Click to view available options
Speed Range:
1400 rpm
1500 rpm
600 rpm
800 rpm
Well Count:
24-well
4-well
8-well
96-well
96/384-well
Tube Size:
0.2 mL
0.5 mL
1.5 mL
2 mL
5 mL
15 mL
50 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Speed Range Well Count Tube Size
14223094 1500 rpm 96/384-well -
14223092 600 rpm 4-well 50 mL
14223089 1500 rpm 8-well 5 mL
14223090 1400 rpm 8-well 5 mL
14223091 800 rpm 8-well 15 mL
14223086 1500 rpm 24-well 0.5 mL
14223087 1500 rpm 24-well 1.5 mL
14223088 1500 rpm 24-well 2 mL
14223093 1500 rpm 96-well 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 14223094 Supplier Axygen™ Supplier No. I4010I
Get Up to $2,500 in Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

These nine interchangeable aluminum blocks accommodate PCR plates and a variety of common life science vessels.

  • No additional tools required for block exchanges
  • Blocks utilize magnetic adhesion technology and are self-recognized once exchanged
  • Combination of heating, cooling, and mixing with versatile block selection makes the Axygen™ Microtube Shaker ideal for many life science laboratories
  • Applications include but are not limited to gene synthesis, gene purification and denaturation, enzymatic reactions, and bacterial growth assays

Specifications

Speed Range 1500 rpm
Material Aluminum
For Use With (Application) PCR
For Use With (Equipment) Axygen™ Microtube Shaker
Well Count 96/384-well