missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Heatex™ Open 1 in. Core, Ink-Side-Out Printer Ribbon - R4300 Wax/Resin, Black

Catalog No. 19104556
Change view
Click to view available options
Color:
Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19104556 Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19104556 Supplier Brady™ Supplier No. 356126

This ink-side-out ribbon features an open 1 in. core for compatibility with a wide variety of thermal transfer printers.

  • One-color ribbon for printing a single color onto a label
  • R4300 Series Ribbons offers strong environmental resistance against smearing and chemicals when printed on the appropriate Brady™ label material
  • Recommended for printing on paper and films that have a matte or rough surface, such as B-427 Self-Laminating Vinyl and B-499 Nylon Cloth from Brady™
  • Combine Brady label materials with their recommended ribbon series for superior legend performance

Specifications

Product Type Printer Ribbon
Color Black
For Use With (Equipment) BradyPrinter PR Plus Series Label Printer, BBP72 Label Printer, Brady X Plus II Series Label Machine, Brady MVP Plus Series, Tagus Thermal Transfer Printer
Length (English) 984 ft.
Width (English) 2.85 in.
Width (Metric) 72.39 mm
Autoclavable Not Autoclavable
Description Printer Ribbon
Format Open-core Ribbon
Quantity 1 Roll