Learn More
Labconco Heated Product Shelf Chambers with Valves
Heated Product Shelf Chambers with Valves for bulk drying and flask drying. Eight ports accommodate both 0.5 and 0.75 in. flask adapters. Includes 12 in. dia. acryl cylind and base.
Supplier:  Labconco 780802100
 View more versions of this product
Catalog No. 17720250
Description
Description
  • Heated Product Shelf Chambers
  • Short (three shelves) or tall (five shelves) options
  • With valves
  • For bulk drying and flask drying
  • 10 in. dia. removable shelves
  • Temperature probes
  • Controlled heat settings
Specifications
Chemical Identifiers
Heated Product Shelf Chamber
-20°C to +50°C
Acrylic
8
Specifications
Heated Product Shelf Chamber
-20°C to +50°C
8
115 V
5
Acrylic
North America
Product Suggestions
Product Suggestions