missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Heated Product Shelf Chambers

Allows drying of bulk samples, or drying of serum bottles, vials or microplates. Labconco Heated Product Shelf Chambers are wireless and removable so that they may be placed in freezer for pre-freezing.

$23433.05 - $32218.29

Specifications

Type Heated Shelf
No. of Shelves 3
Temperature Range (Metric) -20°C to +60°C
Material Steel (Shelves), Acrylic (Chamber)
Certifications/Compliance UL, CAN/CSA, CE
View More Specs

Products 4
Catalog Number Mfr. No. Plug Type Height (Metric) Includes Voltage Price Quantity  
Catalog Number Mfr. No. Plug Type Height (Metric) Includes Voltage Price Quantity  
10400481
View Documents
Labconco™
780801000
North America 43.1 cm 3 shelves and clear chamber 115 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10400482
View Documents
Labconco™
780801010
North America 43.1 cm 3 shelves and clear chamber 230 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10400488
View Documents
Labconco™
780801100
North America 62.4 cm 5 shelves and clear chamber 115 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
10400489
View Documents
Labconco™
780801110
North America 62.4 cm 5 shelves and clear chamber 230 V N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

  • Short Chambers include three 10 in. diameter shelves and three temperature probes; Tall Chambers include five 10 in. diameter shelves and five temperature probes
  • Shelves are wireless and removable so that they may be placed in a freezer for pre-freezing
  • Each shelf is height adjustable to accommodate broad variety of sample sizes
  • Chamber connects to FreeZone Freeze Dryer for control and recording of shelf temperature from Lyo-Works display
  • Temperature may be set from −20° C to 60° C (−4° F to 140° F)
  • Each Chamber includes 12 in. diameter clear acrylic cylinder, vacuum release valve and 8 in. diameter opening to collector on bottom of chamber
  • Will fit 10.12 in. or 14.0 in. diameter condensers
  • For use with any FreeZone Freeze Dryer except Legacy models

Compliance

All models conform to UL Standard 61010-1 (60 Hz models), CAN/CSA C22.2 No. 61010-1 (60 Hz models), and CE Conformity marking (230V, 50 Hz models).

Warranty

One year

Specifications

Specifications

Heated Shelf
-20°C to +60°C
UL, CAN/CSA, CE
38.6 cm
38.6 cm
50/60 Hz
36.29 kg
3
Steel (Shelves), Acrylic (Chamber)
15.2 in.
15.2 in.
2A
80 lb.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'