missing translation for 'onlineSavingsMsg'
Learn More

Mopec™ Heated Forceps System

Catalog No. 22445524
Change view
Click to view available options
Tip Style:
Smooth
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Tip Style
22445524 Smooth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22445524 Supplier Mopec™ Supplier No. BK300SM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The Heated Forceps System is designed to take up minimal bench top space and comes complete with a power supply, control unit, and two different heated forceps.

  • Ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth
  • Choose either serrated or smooth tips
  • System includes: 1 mm Heated Forceps (Red), 2 mm Heated Forceps (Yellow), Mopec™ Control Unit, Mopec™ Power Supply
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth
  • Forceps operate at a temperature between 63°C to 70°C (145°F to 158°F)

Specifications

Temperature (English) 145°F to 158°F
Tip Style Smooth