missing translation for 'onlineSavingsMsg'
Learn More

Supertek™ Heat Conductivity Bar Set

Catalog No. S012331
Encompass
Change view
Click to view available options
Class Size:
1 Lab Group
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S012331 1 Lab Group
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S012331 Supplier Supertek™ Supplier No. PH20272

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A simple, effective apparatus demonstrates the heat conductivity of various metals. Four metal bars (Copper, Aluminum, Brass, and Steel) are mounted on a base with a hole for suspension.

  • Demonstrates the heat conductivity of various metals.
  • Four metal bars (Copper, Aluminum, Brass, and Steel) are mounted on a base with a hole for suspension.
  • Place the bars in a beaker of hot water and compare their relative conductivities.
  • Includes instructions

Specifications

Grade 6+
Includes Instruction Guide
Class Size 1 Lab Group
Product Type Heat Conductometer