missing translation for 'onlineSavingsMsg'
Learn More

Germaine™ Laboratories Healgen™ COVID-19 IgG/IgM Control Kit

Catalog No. 23111398
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23111398 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23111398 Supplier Germaine™ Laboratories Supplier No. LW399

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

An external quality control for anti-SARS-CoV-2 IgM and anti SARS-CoV-2 IgG for use with the COVID-19 IgG/IgM Rapid Test Cassette (whole blood/serum/plasma)

Coronaviruses are enveloped RNA viruses that are distributed broadly among humans, other mammals and birds that cause respiratory, enteric, hepatic and neurologic diseases. Four viruses - 229E, OC43, NL63 and HKU1 are prevalent and typically cause common cold symptoms in immunocompromised individuals. Three other strains SARS-CoV, MERS-CoV and SARS-CoV-2 (COVID-19) are can be transmitted from between non-human vertebrates to humans

  • Use/Purpose:
    • Use with COVID-19 IgG/IgM Rapid Test Cassette (whole blood/serum/plasma) for rapid, qualitative and differential detection of IgG and IgM antibodies to 2019 Novel Coronavirus in human whole blood, serum or plasma to aid in the diagnosis of COVID-19 infection
  • Made in USA

Specifications

Code Color coded caps
Content And Storage -20 °C (12 months); 2°C to 8°C (30 days)
Description IgG/IgM combined control kit
Detectable Analytes anti-SARS-CoV-2 IgM/IgG
For Use With (Equipment) COVID-19 IgG/IgM Rapid Test Cassette
Sample Type Whole Blood, Serum, Plasma
Type Quality Control
Disposable Single-use
Includes 10 total vials - 5 vials of IgG/IgM positive control; 5 vials of negative control. (100 μL / vial).
Quantity 1 Kit
Shipping Condition Overnight services on ice Pk.
Source Human Plasma Based Negative Control. Volume: 0.1 mL.
Sterility Non-sterile
Show More Show Less

PRODUCT MUST BE SHIPPED VIA OVERNIGHT AIR SERVICES ON ICE PACKS.