Learn More
3M™ Headseal, 060-28-03R01
Catalog No. 296357436
Change view
Click to view available options
:
1/Pk.
Catalog No.
296357436 1/Pk.
1 options
Catalog No. 296357436 Supplier 3M™ Supplier No. 7000126345
Only null left
Replacement part for helmets used on 3M™ Breathe Easy Systems, 3M™ Airstream™ Systems, and 3M™ Supplied Air Systems.
  • Powered Air Purifying Respirators are motorized systems powered by a battery pack which draw air through a filter or cartridge to provide respiratory protection to the wearer
  • The blower unit can be mounted on the waist, face, or head, depending on the system selected.
Specifications
Product Type Face Seal
For Use With (Application) For assembly, cleaning and painting
Quantity 1/Pk.