missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Head Activator Neuropeptide

Catalog No. AAJ66870MCR
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66870MCR 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ66870MCR Supplier Thermo Scientific Chemicals Supplier No. J66870MCR

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Purification Method HPLC
Common Name Hydra Peptide
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1125.50
Product Type Head Activator Neuropeptide
Quantity 1 mg
Regulatory Status RUO - research use only
Sequence Glp-Pro-Pro-Gly-Gly-Ser-Lys-Val-Ile-Leu-Phe

RUO – Research Use Only