missing translation for 'onlineSavingsMsg'
Learn More

3M™ HE Filter TR-3712N

Catalog No. p-7155901
Change view
Click to view available options
Quantity:
40/Cs.
5/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19039579 40/Cs.
19039580 5/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19039579 Supplier 3M™ Supplier No. 7100019023
Only null left

HE Filter TR-3712N is a High Efficiency (HE) particulate filter for use with TR-300 Series PAPRs.

  • High Efficiency Particulate Filter
  • Convenient package sizes
  • 3M Catalog# TR-3712N-40 , 3M Catalog# TR-3712N-5

Specifications

Color Magenta
Product Type HE Particulate Filter
For Use With (Equipment) TR-300 Series PAPRs
Model TR-3712N-40
Quantity 40/Cs.