missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Foamed Polyethylene | |
| HDPE | |
| Polypropylene | |
| 61 mm | |
| Scintillation Vial |
| 27 mm | |
| 20 mL | |
| Screw Cap | |
| 1000 Pk. | |
| 22-400 |
| Foamed Polyethylene | |
| HDPE | |
| Polypropylene | |
| 61 mm | |
| Scintillation Vial |
| 27 mm | |
| 20 mL | |
| Screw Cap | |
| 1000 Pk. | |
| 22-400 |