missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ HDPE, Carboy UN/DOT Approved
For Use With: Storage and secure shipments of liquid waste; Closure Material: HDPE; Color: White
$135.96 - $139.11
Specifications
| Certifications/Compliance | UN/DOT |
|---|---|
| Closure Material | HDPE |
| Closure Size | 70 mm |
| Color | White |
| For Use With (Application) | Storage and secure shipments of liquid waste |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|
19-830-450
|
Justrite™
JMC12950 |
13.5 L |
Each for $135.96
|
|
|||||
|
19-830-451
|
Justrite™
JMC12951 |
20 L |
Each for $139.11
|
|
|||||
Description
- For packaging group II and hazardous chemicals
- HDPE construction for excellent chemical compatibility
Specifications
| UN/DOT | |
| 70 mm | |
| Storage and secure shipments of liquid waste | |
| HDPE | |
| Rectangular |
| HDPE | |
| White | |
| 11 in. | |
| Carboy |