missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ HDPE, Carboy
GREENER_CHOICE

Catalog No. 19830409
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
40 L
75 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19830409 5 L
19830408 2.5 L
19830410 10 L
19830413 40 L
19830414 75 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19830409 Supplier Justrite™ Supplier No. 12908
Only null left

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys

Specifications

Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 221 x 132 x 414 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 16.3 in.
Length (English) 8.7 in.
Material HDPE
Product Type Carboy
Shape Rectangular
Width (English) 5.2 in.
Show More Show Less