missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ HDPE, Amber Carboy

For Use With: Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications; Closure Material: Polypropylene; Color: Amber

$271.41 - $623.96

Specifications

Closure Material Polypropylene
Color Amber
For Use With (Application) Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Material HDPE
Product Type Rectangular Carboy
View More Specs

Products 5
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
19-830-420
View Documents
Justrite™
JMC12919
2.5 L
Each for $271.41
Only null left
 
19-830-421
View Documents
Justrite™
JMC12920
5 L
Each for $284.15
Only null left
 
19-830-422
View Documents
Justrite™
JMC12921
10 L
Each for $365.39
Only null left
 
19-830-423
View Documents
Justrite™
JMC12922
20 L
Each for $476.42
Only null left
 
19-830-424
View Documents
Justrite™
12923
40 L
Each for $623.96
Only null left
 
Description

Description

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys
Specifications

Specifications

Polypropylene
Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Rectangular Carboy
Amber
HDPE
Rectangular
Videos
SDS
Documents

Documents