missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ HDPE, Amber Carboy with Spigot

Description
- Interchangeable caps and adapters provide top connection flexibility
- Rectangular ergonomic design
- Material, volume, and metric graduation marks imprinted on carboys
Specifications
Specifications
| Capacity (English) | 1.3 gal. |
| Capacity (Metric) | 5 L |
| Closure Material | Polypropylene |
| Closure Size | 83 mm |
| Color | Amber |
| Dimensions (L x W x H) | 305 x 132 x 414 mm |
| For Use With (Application) | Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications |
| Height (English) | 16.3 in. |
| Length (English) | 12 in. |
| Material | HDPE |
| Show More |