missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ HDPE, Amber Carboy with Spigot
GREENER_CHOICE

Catalog No. 19830425
Change view
Click to view available options
Capacity (Metric):
5 L
10 L
40 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19830425 5 L
19830426 10 L
19830428 40 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19830425 Supplier Justrite™ Supplier No. JMC12924
Only null left

For Use With: Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications; Closure Material: Polypropylene; Color: Amber

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys

Specifications

Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material Polypropylene
Closure Size 83 mm
Color Amber
Dimensions (L x W x H) 305 x 132 x 414 mm
For Use With (Application) Storing and dispensing solvents, buffers, reagents and powders in light sensitive applications
Height (English) 16.3 in.
Length (English) 12 in.
Material HDPE
Product Type Rectangular Carboy
Shape Rectangular
Width (English) 5.2 in.
Show More Show Less