Learn More
Bioassay Systems EnzyChrom™ HDL and LDL/VLDL Assay Kit. For quantitative determination of HDL and LDL/VLDL cholesterol and evaluation of drug effects on HDL and LDL/VLDL metabolism. DFS Item
Supplier:  Bioassay Systems EHDL100
For quantitative determination of HDL and LDL/VLDL cholesterol and evaluation of drug effects on HDL and LDL/VLDL metabolism. Key Features: Sensitive and accurate. Requires only 20 µL serum sample. Detection limit of 5 mg/dL, linearity up to 300 mg/dL cholesterol in 96-well plate assay. Convenient. Room temperature assay. No 37°C heater is needed. Convenient and high-throughput. Homogeneous "mix-incubate-measure" type assay. Can be readily automated on HTS liquid handling systems for processing thousands of samples per day. Method: OD340nm. Samples: Serum. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: 5 mg/dL.
Catalog No. 50489276
Only null left
Explore available promotions