Learn More
Bioassay Systems EnzyChrom™ AF HDL and LDL/VLDL Assay Kit. For quantitative determination of HDL and LDL/VLDL cholesterol and evaluation of drug effects on HDL and LDL/VLDL metabolism.
Supplier:  Bioassay Systems E2HL100
Encompass
For quantitative determination of HDL and LDL/VLDL cholesterol and evaluation of drug effects on HDL and LDL/VLDL metabolism. Key Features: Sensitive and accurate. Linear detection range in 96-well plate: 1 to 100 mg/dL cholesterol for colorimetric assays and 0.2 to 10 mg/dL for fluorimetric assays. Convenient. Room temperature assay. No 37°C heater is needed. Method: OD570nm, or FL530/585nm. Samples: Serum. Species: All. Procedure: Assay takes 60 min. Kit size: 100 tests. Detection limit: OD, FL: 1, 0.2 mg/dL.
Catalog No. 50489238
Only null left