Recombinant human HDAC3 (full-length)/NCOR2 (237-489) complex was expressed by baculovirus in Sf9 insect cells using a C-terminal GST-tag for HDAC3 and N-terminal GST-tag for NCOR2.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human HDAC3 (full-length)/NCOR2 (237-489) complex was expressed by baculovirus in Sf9 insect cells using a C-terminal GST-tag for HDAC3 and N-terminal GST-tag for NCOR2.