Learn More
Lonza Walkersville Inc HCM Hepatocyte Culture Medium BulletKit
Numéro de catalogue. NC9538906
Encompass
The Hepatocyte Culture Medium BulletKit consists of the HBM Hepatocyte Basal Medium, 500 mL (CC-3199) and the HCM SingleQuots Kit (CC-4182).Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for hepatocyte cell healthCompatible with all species hepatocytesContent:1 x HBM (CC-3199) Bottle Basal Medium, 500 mL1 x HCM (CC-4182) Singlequots Supplement Pack containing:1 x Transferrin, 0.50 mL1 x Ascorbic Acid, 0.50 mL1 x HEGF, 0.50 mL1 x Insulin, 0.50 mL1 x Hydrocortisone, 0.50 mL1 x BSA (Fatty Acid Free), 10.5 mL1 x GA-1000, 0.50 mL
Numéro de catalogue. NC9538906 Fournisseur Lonza Walkersville Inc Code fournisseur CC3198
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Hepatocyte Culture Medium BulletKit consists of the HBM Hepatocyte Basal Medium, 500 mL (CC-3199) and the HCM SingleQuots Kit (CC-4182).Once combinded together these reagents formulate a highly reproducible maintenance medium for primary hepatocytes from all species. The final medium is serum-free.Benefits:Optimized for hepatocyte cell healthCompatible with all species hepatocytesContent:1 x HBM (CC-3199) Bottle Basal Medium, 500 mL1 x HCM (CC-4182) Singlequots Supplement Pack containing:1 x Transferrin, 0.50 mL1 x Ascorbic Acid, 0.50 mL1 x HEGF, 0.50 mL1 x Insulin, 0.50 mL1 x Hydrocortisone, 0.50 mL1 x BSA (Fatty Acid Free), 10.5 mL1 x GA-1000, 0.50 mL