Learn More
Innovatek hCG Control Set
Catalog No. 361044577
Change view
Click to view available options
Quantity:
3 Vials/Kit
Catalog No. Quantity
361044577 3 Vials/Kit
1 options
Catalog No. 361044577 Supplier Innovatek Supplier No. 4001QCHCG3
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Innovatek hCG Control Set helps avoid the misinterpretation of pregnancy test results
  • Aids in the interpretation of positive and negative test results and verification of test performance
  • Compatible with all Innovatek brand hCG test kits and other brands of immunochromatograpic kits
  • Unique weak positive trains reader eye to detect the earliest signs of pregnancy avoiding misinterpretation as false negative result
  • Each vial is 4 mL
Specifications
Description 4 mL Vials
Includes Negative, Positive, Weak Positive Controls
Intended Use Pregnancy Test Interpretation
Quantity 3 Vials/Kit