missing translation for 'onlineSavingsMsg'
Learn More

Hourglass International™ HCE™ 2-Ply Cleanroom Face Masks with Earloops

Catalog No. p-4674408
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19423085 Small
19423086 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19423085 Supplier Hourglass International™ Supplier No. FM2ESA14318
Only null left

Cleanroom face masks provide long-wear comfort while protecting the environment from human contaminants including skin flakes, cosmetics, bacteria and other particles. Suitable for Class 100/ISO 5 controlled environments.

  • Low lint design for critical environments
  • 2-ply, white, non-woven ES (PP/PE)/ meltblown polypropylene (BMPP) material
  • Cleanroom manufactured and packed
  • Double bagged cleanroom packaging
  • Soft flexible nose piece
  • Statistical manufacturing process controls
  • OP earloops attached to mask body for strength

Specifications

Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polypropylene
Size Small
Sterility Non-sterile
Style Pleated
Cleanroom Class Class 100 (ISO 5)
Construction 2-ply
Disposable Single-use
For Use With (Application) Semiconductor, Disk Drive, Medical Device, Pharmaceutical, Biomedical
Latex Free Yes
Length (English) 7 in.
Length (Metric) 18 cm
Model FM2ESA143-18
Packaging Quantity 50/Bag, 2000/Cs.
Packaging Type Double-bagged, Cleanroom Packaged
Series HCE™
Show More Show Less