missing translation for 'onlineSavingsMsg'
Learn More

HBTU 98.0+%, TCI America™

Catalog No. B16575G
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16575G 5 g
B165725G 25 g
B1657100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B16575G Supplier TCI America Supplier No. B16575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 94790-37-1
Molecular Formula C11H16F6N5OP
Molecular Weight (g/mol) 379.247
MDL Number MFCD00075445
InChI Key UQYZFNUUOSSNKT-UHFFFAOYSA-N
Synonym hbtu, o-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, 2-1h-benzotriazole-1-yl-1,1,3,3-tetramethyluronium hexafluorophosphate, hbtu-reagent, o-benzotriazol-1-yl-tetramethyluronium hexafluorophosphate, hbtu o-benzotriazole-n,n,n',n'-tetramethyl-uronium-hexafluoro-phosphate, n,n,n',n'-tetramethyl-o-1h-benzotriazol-1-yl uronium hexafluorophosphate, o-1h-benzotriazol-1-yl-n,n,n',n'-tetramethyluronium hexafluorophosphate, o-benzotriazole-n,n,n',n'-tetramethyluronium hexafluorophosphate
PubChem CID 2733084
IUPAC Name [benzotriazol-1-yloxy(dimethylamino)methylidene]-dimethylazanium;hexafluorophosphate
SMILES CN(C)C(=[N+](C)C)ON1C2=CC=CC=C2N=N1.F[P-](F)(F)(F)(F)F

Specifications

Color White
Quantity 5 g
Formula Weight 379.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material HBTU [Coupling Reagent for Peptide]