Learn More
New Pig HazMat Spill Kit in 95 Gallon Wheeled Overpack Salvage Drum
Each absorbs up to 63 gal., absorbs acids, bases, unknowns.
Supplier: New Pig KIT341WH
Description
- Overpack drum container is UN Rated for shipping waste after spill cleanup
- 6 inch olefin wheels make spill kit highly mobile for speedy response anywhere in your facility
- Lift-out, prepacked baskets speed access and guard contents from UV
- PIG HazMat Socks and Dikes stop spreading spills; PIG HazMat Pads and Pillows absorb quickly
- PIG HazMat Absorbents are specially treated for unsurpassed performance with concentrated corrosives, such as 98% sulfuric acid and 30% sodium hydroxide
- Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail
- Lightweight, polyethylene container resists chemicals and keeps contents clean and dry
- Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole
- Ledges molded into container make kit easy to move by forklift
- Tamperproof seals help prevent pilfering of spill response supplies
Specifications
| Polypropylene | |
| Heavyweight | |
| 53.5 kg |
| Overpack Drum Kit | |
| 118 lb. |