missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corepoint™ Scientific Hazardous Location Refrigerator
Delivering professional protection for cold storage in hazardous laboratory locations.
Supplier: Corepoint™ Scientific ER201WWW0HC
Description
Hazardous location refrigerators meet NFPA and OSHA guidelines for Hazardous Location, Class 1, Division II, Groups C and D guidelines.
- These units utilize non-sparking and/or ATEX rated components and are engineered for use in environments where volatile/explosive conditions could potentially exist.
- Models feature a microprocessor temperature controller, allowing for precise temperature control, recovery and maintenance versus mechanical style thermostats.
- Our refrigerators run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
- For storage of flammable or volatile materials within typical laboratory or non-volatile environments, see Flammable Material Storage units.
Specifications
| HazLoc Refrigerator | |
| 566 L | |
| Solid Swing | |
| 5 | |
| Powder-coated Steel | |
| Upright | |
| H-drive | |
| 115 V |
| 20 cu. ft. | |
| 1°C to 10°C | |
| 1 | |
| Scratch-resistant Painted Steel | |
| cULus, ETL Listed, ENERGYSTAR | |
| Microprocessor Temperature Controller | |
| No Plug, Hardwired |