missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ T, 100-120 mesh

Catalog No. 111004202
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004202 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004202 Supplier MilliporeSigma™ Supelco™ Supplier No. 10311

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

HayeSep™ T was used together with a thermoconductivity detector to study the solubility of trioxane in methanol at different temperatures and to measure the vapour pressure data for this binary mixture.

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep™ has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity. HayeSep™ T is a copolymer of ethylene glycol methacrylate (EGDMA).

Specifications

Particle Size 100/120 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜250 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.38 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.