missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ D, 100-120 mesh
Description
HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.
Specifications
Specifications
| Particle Size | 100/120 Mesh |
| Packaging | Bottle |
| Quantity | 75 cc |
| Surface Area (Metric) | ˜795 m2/g |
| Product Line | HayeSep |
| For Use With (Application) | GC |
| Density | ˜0.33 g/mL (free fall density) |
HayeSep is a registered trademark of Hayes Separation Inc.