missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ C, 80-100 mesh

Catalog No. 111004188
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004188 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004188 Supplier MilliporeSigma™ Supelco™ Supplier No. 10289

HayeSep™ C maybe used in an experimental set-up to remove NH3 from H2SO4 aqueous solution for hydrodenitrogenation of quinolone.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜442 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.32 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.