missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ C, 100-120 mesh
Description
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. HayeSep™ has a high surface area owing to its micropores thereby making it a suitable candidate for separation of gases and volatile organic compounds (VOCs). Furthermore they are relatively inert and exhibit hydrophobicity. HayeSep™ C is a copolymer of divinylbenzene and acrylonitrile.
Specifications
Specifications
| Particle Size | 100/120 Mesh |
| Packaging | Bottle |
| Quantity | 75 cc |
| Surface Area (Metric) | ˜442 m2/g |
| Product Line | HayeSep |
| For Use With (Application) | GC |
| Density | ˜0.32 g/mL (free fall density) |
HayeSep is a registered trademark of Hayes Separation Inc.