missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ B, 80-100 mesh

Catalog No. 111004186
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004186 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004186 Supplier MilliporeSigma™ Supelco™ Supplier No. 10286

HayeSep™ B can maybe used in combination with HayeSep™ A for separation of a gaseous mixture comprising of hydrogen and hydrogen sulphide.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜608 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.33 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.