missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ HayeSep™ Porous Polymer Adsorbent, Matrix HayeSep™ A, 80-100 mesh

Catalog No. 111004184
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004184 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004184 Supplier MilliporeSigma™ Supelco™ Supplier No. 10283

HayeSep™A maybe used for the measurement of carbon dioxide using the on-line gas analyser during the experimental set-up, to use activated carbon as a buffer in biofiltration of waste gases, to ensure a constant supply of contaminants like toluene to the bioreactor for effective biological waste-gas treatment.

HayeSep™ porous polymers, considered second generation materials, are consistent batch-to-batch, with minimal shrinkage and monomer bleed.

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) 526 m2/g
Product Line HayeSep
For Use With (Application) GC
Density ˜0.36 g/mL (free fall density)

HayeSep is a registered trademark of Hayes Separation Inc.