Learn More
Haws™ Thermal Protection Jackets
Model 9125 is a white thermal protection jacket for 7500 series gravity eyewashes.
Supplier:  Haws™ 9125
Catalog No. 29663195
Only null left
Description
Description
Model 9125 is a thermal protection jacket. Recommended for areas where extreme heat or sunlight might cause water to overheat
Specifications
Specifications
Thermal Protection Jacket
Product Suggestions
Product Suggestions