missing translation for 'onlineSavingsMsg'
Learn More

Haws™ Shower Test Kit

Catalog No. 1798555
Change view
Click to view available options
For Use With (Equipment):
8130, 8133H, Emergency Shower
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
1798555 8130, 8133H, Emergency Shower
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1798555 Supplier Haws™ Supplier No. HAWS9010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Haws™ Shower Test Kit is for use with emergency showers

Model 9010 shower test kit affords periodic shower testing with ease. The kit includes an upper corrosion resistant ring to encircle the shower head and tunnel shaped yellow Tyvek waterproof tube to guide the water column to model 9009 waste water container, or drain. Holding rod can be used as a handle for ease of testing by user. Testing record card included.

Warranty

1 year from ship date (Limited)

Specifications

Color Yellow
For Use With (Equipment) 8130, 8133H, Emergency Shower
Height (English) 78 in.
Height (Metric) 198.12 cm
Material Plastic
Product Type Shower Test Kit