Learn More
Haws™ Portable Eyewash Cart
Model 9008 cart designed to transport all stainless steel portable eyewashes.
Supplier: Haws™ 9008
Description
Model 9008 cart designed to transport all stainless steel portable eyewashes. Unit is constructed of heavy-duty 1 in. (2.5 cm) tubular frame with green powder-coated finish, features rubber wheels, and is equipped with a ratchet tie down strap to securely hold the tank to the cart.
Spécifications
| Ratchet Tie Down Strap | |
| Stainless Steel | |
| Portable Eyewash Cart |