missing translation for 'onlineSavingsMsg'
Learn More

Haws™ Eye/Face Wash Head Assemblies

Catalog No. 19039691
Change view
Click to view available options
Includes:
Built-in 2.5 gpm Flow Control, Integral Flip Top Dust Cover
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
19039691 Built-in 2.5 gpm Flow Control, Integral Flip Top Dust Cover
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19039691 Supplier Haws™ Supplier No. SP21FC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Assures multiple soft-and-steady streams of water

  • Flowing pressure range from 2.1 to 6.2 bar (30 to 90 psi)
  • Top dust cover that automatically releases with water flow
  • Built-in 2.5 gpm flow control
  • Green plastic

Specifications

Color Green
Includes Built-in 2.5 gpm Flow Control, Integral Flip Top Dust Cover
Material ABS Plastic
Product Type Head Assembly