missing translation for 'onlineSavingsMsg'
Learn More

Haws™ Emergency Shower and Eyewash Receptors

Catalog No. 19811871
Change view
Click to view available options
Material:
ABS Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19811871 ABS Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19811871 Supplier Haws™ Supplier No. SP93

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use on most Eyewashes and Eye/Face. Wash Stations series

Model SP93 is a Safety Green 11 in. (27.9 cm) ABS plastic receptor resistant to damage from alkalies, salt solutions, oils and most acids. For use on most Eyewashes and Eye/Face. Wash Stations series

Specifications

Color Green
For Use With (Equipment) Shower, Eyewash
Height (English) 3.56 in.
Height (Metric) 9.04 cm
Material ABS Plastic
Product Type Receptor