Learn More
Hausser Scientific™ Cover Glasses/Slips
Crafted from high-quality borosilicate glass.
Supplier:  Hausser Scientific™ 5051
 View more versions of this product
Catalog No. 02617582
Only null left
Description
Description
Crafted from high-quality borosilicate glass
Specifications
Chemical Identifiers
Coverslip
Rectangle
Borosilicate Glass
1 Ea.
Specifications
Coverslip
Borosilicate Glass
26 mm
Petroff-Hausser Counter
Rectangle
20 mm
0.2 mm
1 Ea.
Product Suggestions
Product Suggestions