Learn More
Hausser Scientific™ Bright-Line™ Counting Chamber
Description
Diamond etched with double improved Neubauer ruling
- One piece construction, ensuring long term durability and accuracy
- 'V' slash at the loading side of each plateau facilitates charing and reduces the possibility of overflow into the moat
- Each plateau features enhanced Neubauer rulings
- Ruled surface is 0.1mm below the cover glass (sold separately), limiting the volume of blood or fluid over a square mm at 0.1cu.mm and over each of 400 squares (within the central square mm) to 0.00025 cu. Mm.
- Contact of the flat, polished cover glass surfaces with cover glass supports produces an exact volume of fluid over the counting area
- Difference in surface tension characteristics between the metallic surface on the chamber and the polished cover glass assures smooth capillarity for precise loading and more even cell distribution
Specifications
Specifications
| Product Type | Hemacytometer |
| Grid Type | Bright Line |
| Capacity (Metric) | 0.1μL |
| Material | Milled Soda Lime Glass |
| Quantity | 1 Ea. |
| For Use With (Application) | Double Improved Neubauer Ruling |