Learn More
Hausser Scientific™ Hy-Lite™ Counting Chamber Set
Supplied with polyethylene carrying case
Includes: Counting chamber (02-671-10), two cover glasses, two Thoma pipets (one for red and one for white corpuscles), and pipet filler.
Description
Supplied with polyethylene carrying case
Specifications
| Hemacytometer Chamber Set | |
| 0.1μL | |
| 0.003 in. | |
| Consistent Cell Counting |
| Hausser Hy-Lite | |
| Each | |
| 0.01cm | |
| Counting chamber, two cover Glasses, two pipets (one for red and one for white corpuscles) and pipet filler, Polyethylene carrying case |