missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Hatu

Catalog No. 44546025G
Encompass
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44546025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44546025G Supplier Sigma Aldrich Supplier No. 44546025G

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hatu

Specifications

CAS 148893-10-1
Melting Point 183°C to 185°C
Molecular Formula C10H15F6N6OP
Linear Formula C10H15F6N6OP
Quantity 25 g
MDL Number MFCD27957364
Synonym 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-oxid hexafluorophosphate; N-[(Dimethylamino)-1H-1,2,3-triazolo-[4,5-b]pyridin-1-ylmethylene]-N-methylmethanaminium hexafluorophosphate N-oxide
Molecular Weight (g/mol) 380.23 g/mol