Learn More
Thermo Scientific™ Hastelloy PT1000 External Probe
Catalog No. CIC0001449
Change view
Click to view available options
Type:
Probe
Catalog No. Type
CIC0001449 Probe
1 options
Catalog No. CIC0001449 Supplier Thermo Scientific™ Supplier No. CIC0001449
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
For Use With: Cimarec and Supernuova Hotplates and Hotplate Stirrers
Specifications
Description Hastelloy PT1000 External probe, 0-400°C, Hastelloy, for all sizes of Cimarec+ and SuperNuova+ Hotplates and Hotplate Stirrers
For Use With (Equipment) Cimarec and Supernuova Hotplates and Hotplate Stirrers
Temperature (Metric) 0°C to 400°C
Type Probe
Display Digital