missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hastelloy C∣r gauze, 100 mesh woven from 0.102mm (0.004in) dia wire
$233.01 - $233.01
Specifications
| Chemical Name or Material | Hastelloy C™ gauze |
|---|---|
| Name Note | Woven from 0.102mm (0.004 in.) dia. wire |
| Mesh Size | 100 Mesh |
| Form | Wire Cloth |
| MDL Number | MFCD00801065 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Hastelloy C™ gauze | |
| 100 Mesh | |
| MFCD00801065 |
| Woven from 0.102mm (0.004 in.) dia. wire | |
| Wire Cloth |
RUO – Research Use Only