missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Harpagoside, ≥95% (HPLC), MilliporeSigma™ Supelco™
Description
Harpagoside, a naturally occurring iridoid glycoside, is the main bioactive component of medicinal plants like Radix Scrophulariae, Scrophularia ningpoensis, Scrophularia buergeriana and Harpagophytum procumbens. It can exhibit a wide range of biological and pharmacological properties and hence used in the treatment of ailments like pharyngalgia, arthritis, rheumatism, tussis and constipation. Featured Industries: Food and Beverages
Specifications
Specifications
| CAS | 19210-12-9 |
| Physical Form | Neat |
| Linear Formula | C24H30O11 |
| MDL Number | MFCD00017415 |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 494.49 |
| Percent Purity | ≥95% (HPLC) |
| Grade | Analytical Standard |